Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024363.1 | NP_077339.1 | 7 - 8 | 708 | 123 | |
XM_006229144.2 | XP_006229206.1 | 8 - 9 | 826 | 123 | ||
XM_006229145.2 | XP_006229207.1 | 7 - 8 | 990 | 123 | ||
XM_006229146.2 | XP_006229208.1 | 8 - 9 | 1093 | 123 | ||
GenBank mRNA | BC078815.1 | - | 7 - 8 | 713 | 7106257 | 123 |
FQ213198.1 | - | 7 - 8 | 721 | 123 | ||
FQ219843.1 | - | 7 - 8 | 719 | 123 | ||
FQ220982.1 | - | 5 - 6 | 460 | 123 | ||
FQ228625.1 | - | 7 - 8 | 989 | 123 | ||
U60882.1 | - | 7 - 8 | 708 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |