Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031840.1 | NP_114028.1 | 6 - 7 | 884 | 88 | |
GenBank mRNA | BC059125.1 | - | 6 - 7 | 812 | 6919326 | 88 |
FQ209697.1 | - | 6 - 7 | 777 | 88 | ||
FQ210400.1 | - | 6 - 7 | 776 | 88 | ||
FQ211073.1 | - | 7 - 8 | 993 | 88 | ||
FQ213475.1 | - | 6 - 7 | 808 | 88 | ||
FQ218913.1 | - | 6 - 7 | 779 | 88 | ||
FQ218933.1 | - | 6 - 7 | 778 | 88 | ||
FQ219092.1 | - | 6 - 7 | 778 | 88 | ||
FQ225110.1 | - | 6 - 7 | 846 | 88 | ||
M17300.1 | - | 5 - 6 | 754 | 88 | ||
M34477.1 | - | 6 - 7 | 884 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |