Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053467.1 | NP_445919.1 | 1 - 2 | 224 | 99 | |
GenBank mRNA | AJ004912.1 | - | 1 - 2 | 224 | 99 | |
X97443.1 | - | 1 - 2 | 176 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |