Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257671.2 | XP_006257733.1 | 5 - 6 | 938 | 61 | |
XM_006257672.2 | XP_006257734.1 | 6 - 7 | 874 | 61 | ||
XM_006257673.2 | XP_006257735.1 | 6 - 7 | 897 | 61 | ||
XM_006257674.2 | XP_006257736.1 | 6 - 7 | 891 | 61 | ||
XM_006257675.2 | XP_006257737.1 | 5 - 6 | 847 | 61 | ||
XM_006257676.2 | XP_006257738.1 | 5 - 6 | 817 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |