Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100542.1 | NP_001094012.1 | 6 - 7 | 659 | 148 | |
GenBank mRNA | BC091286.1 | - | 4 - 5 | 442 | 148 | |
BC166783.1 | - | 6 - 7 | 641 | 7935021 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |