Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109029.1 | NP_001102499.1 | 2 - 3 | 463 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |