Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acyp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106746.2 | NP_001100216.1 | 2 - 3 | 237 | 99 | |
NM_001199167.1 | NP_001186096.1 | 2 - 3 | 169 | 99 | ||
NM_001199168.1 | NP_001186097.1 | 2 - 3 | 133 | 99 | ||
XM_006240308.2 | XP_006240370.1 | 2 - 3 | 288 | 99 | ||
GenBank mRNA | FM034312.1 | - | 2 - 3 | 133 | 99 | |
FM062949.1 | - | 2 - 3 | 225 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |