Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047116.1 | NP_001040581.1 | 3 - 4 | 612 | 164 | |
XM_006236003.2 | XP_006236065.1 | 3 - 4 | 1656 | 164 | ||
GenBank mRNA | BC107563.1 | - | 3 - 4 | 612 | 7938579 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |