Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109541.1 | NP_001103011.1 | 7 - 8 | 772 | 60 | |
XM_006245276.1 | XP_006245338.1 | 8 - 9 | 831 | 60 | ||
XM_006245278.2 | XP_006245340.1 | 8 - 9 | 853 | 60 | ||
XM_006245279.2 | XP_006245341.1 | 8 - 9 | 780 | 60 | ||
XM_006245280.1 | XP_006245342.1 | 8 - 9 | 831 | 60 | ||
XM_006245281.2 | XP_006245343.1 | 7 - 8 | 780 | 60 | ||
XM_006245283.1 | XP_006245345.1 | 8 - 9 | 831 | 60 | ||
GenBank mRNA | BC166710.1 | - | 8 - 9 | 768 | 7930287 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |