Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134750.1 | NP_001128222.1 | 6 - 7 | 621 | 62 | |
NM_001134751.1 | NP_001128223.1 | 5 - 6 | 539 | 62 | ||
GenBank mRNA | BC168228.1 | - | 6 - 7 | 621 | 7374541 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |