Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107236.1 | NP_001100706.1 | 3 - 4 | 646 | 131 | |
XM_008770308.1 | XP_008768530.1 | 3 - 4 | 1924 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |