Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012819.1 | NP_036951.1 | 5 - 6 | 610 | 68 | |
GenBank mRNA | BC062006.1 | - | 5 - 6 | 608 | 5599828 | 68 |
FQ215575.1 | - | 5 - 6 | 661 | 68 | ||
J05029.1 | - | 5 - 6 | 621 | 68 | ||
L11276.1 | - | 5 - 6 | 610 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |