Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024143.2 | NP_077057.1 | 7 - 8 | 1716 | 60 | |
GenBank mRNA | AF242189.1 | - | 7 - 8 | 1717 | 60 | |
BC091147.1 | - | 7 - 8 | 1739 | 7370589 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |