Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033924.1 | NP_001029096.1 | 3 - 4 | 259 | 58 | |
NM_133550.2 | NP_598234.2 | 3 - 4 | 259 | 58 | ||
XM_008768939.1 | XP_008767161.1 | 2 - 3 | 423 | 58 | ||
GenBank mRNA | AF381978.1 | - | 2 - 3 | 145 | 58 | |
BC078749.1 | - | 3 - 4 | 259 | 7124300 | 58 | |
X73579.1 | - | 2 - 3 | 142 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |