Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014094.1 | NP_001014116.1 | 1 - 2 | 52 | 68 | |
XM_008766450.1 | XP_008764672.1 | 1 - 2 | 300 | 68 | ||
GenBank mRNA | BC079090.1 | - | 1 - 2 | 52 | 7123519 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |