Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244025.2 | XP_006244087.1 | 9 - 10 | 1945 | 83 | |
XM_008766657.1 | XP_008764879.1 | 9 - 10 | 2002 | 83 | ||
XM_243980.7 | XP_243980.4 | 9 - 10 | 3097 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |