Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012146.1 | NP_001012146.1 | 2 - 3 | 477 | 84 | |
XM_008768890.1 | XP_008767112.1 | 2 - 3 | 451 | 84 | ||
GenBank mRNA | BC081905.1 | - | 2 - 3 | 477 | 7111034 | 84 |
FQ211720.1 | - | 2 - 3 | 401 | 84 | ||
FQ228410.1 | - | 2 - 3 | 379 | 84 |
Gene Symbol: LOC100910275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750241.2 | XP_003750289.1 | 2 - 3 | 476 | 84 | |
XM_006240675.2 | XP_006240737.1 | 2 - 3 | 444 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |