Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107079.1 | NP_001100549.1 | 2 - 3 | 287 | 73 | |
XM_006248699.2 | XP_006248761.1 | 2 - 3 | 629 | 73 | ||
XM_006248700.2 | XP_006248762.1 | 2 - 3 | 629 | 73 | ||
XM_006248701.2 | XP_006248763.1 | 3 - 4 | 516 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |