Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106155.1 | NP_001099625.1 | 1 - 2 | 122 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |