Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chaf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024741.1 | NP_001019912.1 | 5 - 6 | 598 | 91 | |
XM_006248044.2 | XP_006248106.1 | 5 - 6 | 634 | 91 | ||
GenBank mRNA | BC097460.1 | - | 5 - 6 | 598 | 7457454 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |