Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034933.2 | NP_001030105.2 | 4 - 5 | 851 | 67 | |
GenBank mRNA | BC105852.1 | - | 4 - 5 | 1232 | 7452956 | 67 |
FQ212178.1 | - | 1813 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |