Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024877.1 | NP_001020048.1 | 7 - 8 | 644 | 65 | |
XM_006242186.2 | XP_006242248.1 | 6 - 7 | 606 | 65 | ||
XM_006242187.2 | XP_006242249.1 | 6 - 7 | 606 | 65 | ||
XM_006242188.2 | XP_006242250.1 | 6 - 7 | 567 | 65 | ||
GenBank mRNA | BC097379.1 | - | 7 - 8 | 644 | 7445599 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |