Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108731.1 | NP_001102201.1 | 6 - 7 | 901 | 114 | |
XM_006240944.2 | XP_006241006.1 | 7 - 8 | 798 | 114 | ||
XM_006240946.2 | XP_006241008.1 | 7 - 8 | 720 | 114 | ||
GenBank mRNA | FQ220489.1 | - | 2 - 3 | 106 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |