Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012114.2 | NP_001012114.2 | 9 - 10 | 2965 | 72 | |
GenBank mRNA | BC083596.1 | - | 5 - 6 | 960 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |