Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108072.2 | NP_001101542.1 | 3 - 4 | 271 | 90 | |
XM_006240928.1 | XP_006240990.1 | 3 - 4 | 298 | 90 | ||
XM_006240929.1 | XP_006240991.1 | 3 - 4 | 298 | 90 | ||
XM_006240930.1 | XP_006240992.1 | 3 - 4 | 298 | 90 | ||
GenBank mRNA | BC168179.1 | - | 3 - 4 | 271 | 9092463 | 90 |
FQ230285.1 | - | 3 - 4 | 540 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |