Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014168.1 | NP_001014190.1 | 4 - 5 | 613 | 97 | |
XM_006231545.2 | XP_006231607.1 | 3 - 4 | 519 | 97 | ||
GenBank mRNA | BC085841.1 | - | 4 - 5 | 613 | 7135812 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |