Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231682.1 | XP_006231744.1 | 1 - 2 | 109 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |