Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126093.2 | NP_001119565.1 | 1 - 2 | 199 | 115 | |
GenBank mRNA | BC086330.1 | - | 1 - 2 | 199 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |