Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109079.2 | NP_001102549.2 | 7 - 8 | 2094 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |