Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_223012.8 | XP_223012.6 | 2 - 3 | 412 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Rattus norvegicus Probe spans exons |