Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014787.1 | NP_001014787.1 | 4 - 5 | 976 | 62 | |
XM_006250286.2 | XP_006250348.1 | 4 - 5 | 1135 | 62 | ||
XM_006250288.2 | XP_006250350.1 | 3 - 4 | 1077 | 62 | ||
XM_006250289.2 | XP_006250351.1 | 3 - 4 | 914 | 62 | ||
GenBank mRNA | BC092192.1 | - | 4 - 5 | 976 | 7312924 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |