Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106005.1 | NP_001099475.1 | 4 - 5 | 534 | 107 | |
GenBank mRNA | BC168866.1 | - | 4 - 5 | 521 | 7460205 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |