Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022869.1 | NP_074060.1 | 9 - 10 | 1181 | 86 | |
XM_006231560.2 | XP_006231622.1 | 9 - 10 | 1229 | 86 | ||
GenBank mRNA | M94287.1 | - | 9 - 10 | 1181 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |