Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025671.1 | NP_001020842.1 | 3 - 4 | 330 | 67 | |
XM_006250745.2 | XP_006250807.1 | 4 - 5 | 442 | 67 | ||
XM_006250747.2 | XP_006250809.1 | 4 - 5 | 430 | 67 | ||
XM_006250748.1 | XP_006250810.1 | 4 - 5 | 465 | 67 | ||
XM_008770036.1 | XP_008768258.1 | 4 - 5 | 525 | 67 | ||
XM_008770037.1 | XP_008768259.1 | 4 - 5 | 475 | 67 | ||
XM_008770038.1 | XP_008768260.1 | 4 - 5 | 3717 | 67 | ||
GenBank mRNA | BC097375.1 | - | 3 - 4 | 330 | 7444629 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |