Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025141.1 | NP_001020312.1 | 1 - 2 | 185 | 99 | |
XM_006251901.2 | XP_006251963.1 | 2 - 3 | 595 | 99 | ||
XM_006251903.2 | XP_006251965.1 | 1 - 2 | 698 | 99 | ||
XM_008770563.1 | XP_008768785.1 | 1 - 2 | 699 | 99 | ||
XM_008770564.1 | XP_008768786.1 | 2 - 3 | 596 | 99 | ||
XM_008770565.1 | XP_008768787.1 | 2 - 3 | 469 | 99 | ||
XM_008770566.1 | XP_008768788.1 | 1 - 2 | 699 | 99 | ||
XM_008770567.1 | XP_008768789.1 | 1 - 2 | 698 | 99 | ||
XM_008770568.1 | XP_008768790.1 | 1 - 2 | 698 | 99 | ||
GenBank mRNA | BC091423.1 | - | 1 - 2 | 185 | 7365771 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |