Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024794.1 | NP_001019965.1 | 1 - 2 | 164 | 104 | |
XM_008770555.1 | XP_008768777.1 | 1 - 2 | 537 | 104 | ||
GenBank mRNA | BC097400.1 | - | 1 - 2 | 164 | 7453630 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |