Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250443.2 | XP_006250505.1 | 1 - 2 | 996 | 90 | |
GenBank mRNA | AB126962.1 | - | 1 - 2 | 227 | 90 | |
AY341057.1 | - | 1 - 2 | 106 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Nuclear Receptor Rattus norvegicus Probe spans exons |