Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109343.1 | NP_001102813.1 | 3 - 4 | 583 | 113 | |
XM_006235162.2 | XP_006235224.1 | 4 - 5 | 1174 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |