Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133608.2 | NP_598292.2 | 7 - 8 | 1641 | 59 | |
GenBank mRNA | AF239262.1 | - | 6 - 7 | 1475 | 59 | |
BC097285.1 | - | 7 - 8 | 1641 | 7441748 | 59 | |
BC097286.1 | - | 7 - 8 | 1583 | 7444297 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |