Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191967.1 | NP_001178896.1 | 3 - 4 | 448 | 65 | |
XM_006235798.1 | XP_006235860.1 | 3 - 4 | 459 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |