Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 8 - 9 | 834 | 66 | |
NM_134472.3 | NP_608302.2 | 7 - 8 | 719 | 66 | ||
XM_006238632.2 | XP_006238694.1 | 8 - 9 | 886 | 66 | ||
XM_006238633.1 | XP_006238695.1 | 7 - 8 | 650 | 66 | ||
XM_006238634.2 | XP_006238696.1 | 7 - 8 | 1182 | 66 | ||
XM_006238635.2 | XP_006238697.1 | 8 - 9 | 1059 | 66 | ||
XM_006238636.2 | XP_006238698.1 | 8 - 9 | 1055 | 66 | ||
GenBank mRNA | BF289656.1 | - | 1 - 2 | 53 | 66 | |
CO575506.1 | - | 7 - 8 | 642 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |