Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238579.2 | XP_006238641.1 | 2 - 3 | 164 | 65 | |
XM_345561.7 | XP_345562.4 | 2 - 3 | 175 | 65 | ||
GenBank mRNA | FQ222511.1 | - | 1 - 2 | 46 | 65 | |
FQ222777.1 | - | 2 - 3 | 172 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |