Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108153.1 | NP_001101623.1 | 2 - 3 | 326 | 59 | |
XM_008766273.1 | XP_008764495.1 | 2 - 3 | 173 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |