Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024801.1 | NP_001019972.1 | 9 - 10 | 1353 | 79 | |
GenBank mRNA | BC097452.1 | - | 9 - 10 | 1353 | 7456845 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |