Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb41l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053927.1 | NP_446379.1 | 6 - 7 | 696 | 78 | |
XM_006245649.2 | XP_006245711.1 | 6 - 7 | 3063 | 78 | ||
XM_006245651.2 | XP_006245713.1 | 6 - 7 | 3057 | 78 | ||
XM_006245652.2 | XP_006245714.1 | 6 - 7 | 3058 | 78 | ||
XM_008767414.1 | XP_008765636.1 | 6 - 7 | 3069 | 78 | ||
XM_008767415.1 | XP_008765637.1 | 6 - 7 | 3056 | 78 | ||
XM_008767416.1 | XP_008765638.1 | 6 - 7 | 3056 | 78 | ||
XM_008767417.1 | XP_008765639.1 | 6 - 7 | 3056 | 78 | ||
GenBank mRNA | AB032827.1 | - | 6 - 7 | 696 | 78 | |
AB032828.1 | - | 6 - 7 | 1137 | 78 | ||
DQ462202.1 | - | 5 - 6 | 616 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |