Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252959.2 | XP_006253021.1 | 2 - 3 | 290 | 93 | |
XM_006252960.2 | XP_006253022.1 | 2 - 3 | 289 | 93 | ||
XM_006252961.2 | XP_006253023.1 | 2 - 3 | 288 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |