Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107304.1 | NP_001100774.1 | 9 - 10 | 1488 | 70 | |
XM_008771106.1 | XP_008769328.1 | 9 - 10 | 1375 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |