Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271055.1 | NP_001257984.1 | 7 - 8 | 935 | 120 | |
XM_006252897.2 | XP_006252959.1 | 8 - 9 | 1005 | 120 | ||
GenBank mRNA | BC089056.1 | - | 7 - 8 | 862 | 120 | |
FQ212329.1 | - | 7 - 8 | 910 | 120 | ||
FQ214027.1 | - | 7 - 8 | 890 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |