Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030026.1 | NP_001025197.1 | 1 - 2 | 254 | 131 | |
GenBank mRNA | BC088256.1 | - | 1 - 2 | 118 | 131 | |
BC099774.1 | - | 1 - 2 | 254 | 7932222 | 131 | |
FQ225096.1 | - | 1 - 2 | 140 | 131 | ||
FQ229307.1 | - | 1 - 2 | 292 | 131 | ||
FQ230297.1 | - | 1 - 2 | 140 | 131 | ||
FQ232607.1 | - | 1 - 2 | 140 | 131 | ||
FQ233058.1 | - | 1 - 2 | 140 | 131 | ||
FQ233063.1 | - | 1 - 2 | 172 | 131 | ||
FQ233214.1 | - | 1 - 2 | 140 | 131 | ||
FQ234315.1 | - | 1 - 2 | 140 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |