Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108398.1 | NP_001101868.1 | 2 - 3 | 211 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |